missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BTN2A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562421
This item is not returnable.
View return policy
Description
Immunogen sequence: EKKVEQEEKE IAQQLQEELR Highest antigen sequence identity to the following orthologs: Mouse - 75%, Rat - 65%.
Butyrophilin is the major protein associated with fat droplets in the milk. This gene is a member of the BTN2 subfamily of genes, which encode proteins belonging to the butyrophilin protein family. The gene is located in a cluster on chromosome 6, consisting of seven genes belonging to the expanding B7/butyrophilin-like group, a subset of the immunoglobulin gene superfamily. The encoded protein is a type I receptor glycoprotein involved in lipid, fatty-acid and sterol metabolism. Several alternatively spliced transcript variants encoding different isoforms have been found for this gene.
Specifications
| BTN2A2 | |
| Polyclonal | |
| Unconjugated | |
| BTN2A2 | |
| BT2.2; BTF2; Btn2; BTN2.2; BTN2A2; butyrophilin 2; butyrophilin subfamily 2 member A2; butyrophilin, subfamily 2, member A2; D030063K05 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 10385 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WVV5 | |
| BTN2A2 | |
| Recombinant protein corresponding to Human BTN2A2. Recombinant protein control fragment (Product #RP-101302). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction