Learn More
BTN2A1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA587883
Description
Immunogen sequence: FIVVGPTDPI LATVGENTTL RCHLSPEKNA EDMEVRWFRS QFSPAVFVYK GGRERTEEQM EEYRGRTTFV SKDISRGSVA LVIHNITAQE NGTYRCYFQE GRSYDEAILH LVVAGLGSKP LISMRGHEDG GIRLECISRG WYPKPLTVWR DPYGGVAPAL KEVSMPDADG LFMVTTAVII RDKSVRNMSC SINNTLLGQK KESVIFIPES F; Positive Samples: U-251MG, HT-1080, HT-29, LO2, A-549, 293T; Cellular Location: Membrane, Single-pass type I membrane protein
The specific functin of this protein remains unknown.This gene is a member of the BTN2 subfamily of genes, which encode proteins belonging to the butyrophilin protein family. The gene is located in a cluster on chromosome 6, consisting of seven genes belonging to the expanding B7/butyrophilin-like group, a subset of the immunoglobulin gene superfamily. The encoded protein is an integral plasma membrane B box protein involved in lipid, fatty-acid and sterol metabolism.
Specifications
| BTN2A1 | |
| Polyclonal | |
| Unconjugated | |
| BTN2A1 | |
| BTN2A1 | |
| Recombit fusion protein containing a sequence corresponding to amino acids 30-240 of human BTN2A1 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| 4.52 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q7KYR7 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 11120 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.