missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTN2A1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£328.00 - £443.00
Specifications
| Antigen | BTN2A1 |
|---|---|
| Applications | Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18462121
|
Novus Biologicals
NBP1-88504-25ul |
25 μL |
£328.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18226078
|
Novus Biologicals
NBP1-88504 |
0.1 mL |
£443.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BTN2A1 Polyclonal specifically detects BTN2A1 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.Specifications
| BTN2A1 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| BK14H9.1, BTF1BT2.1butyrophilin BTF1, butyrophilin subfamily 2 member A1, butyrophilin, subfamily 2, member A1, DJ3E1.1, FLJ36567 | |
| BTN2A1 | |
| IgG | |
| Affinity Purified | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| 11120 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:ILSGEKEFERETREIALKELEKERVQKEEELQVKEKLQEELRWRRTFLHAVDVVLD | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title