missing translation for 'onlineSavingsMsg'
Learn More

BTN2A1 Antibody (ICT-01-N297A) [Janelia Fluor™ 635], Novus Biologicals™

Product Code. 30162897
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30162897 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30162897 Supplier Novus Biologicals (Bio-Techne) Supplier No. NBP328457JF635

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Human Monoclonal Antibody

BTN2A1 Monoclonal antibody specifically detects BTN2A1 in Human samples. It is validated for Flow Cytometry,ELISA,Functional Assay
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTN2A1
Applications Flow Cytometry, ELISA, Functional Assay
Classification Monoclonal
Clone ICT-01-N297A
Conjugate Janelia Fluor 635
Formulation 50mM Sodium Borate
Gene Alias BK14H9.1, BTF1BT2.1butyrophilin BTF1, butyrophilin subfamily 2 member A1, butyrophilin, subfamily 2, member A1, DJ3E1.1, FLJ36567
Host Species Human
Immunogen BTN1A1
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Endocrinology, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 11120
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.