missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BTLA Monoclonal Antibody (K06312_3D7), Invitrogen™

Mouse Monoclonal Antibody

£416.00 - £734.00

Specifications

Antigen BTLA
Clone K06312_3D7
Concentration 1 mg/mL
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
30205618
View Documents
Invitrogen™
MA555989
50 μL
£416.00
50µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
30203665
View Documents
Invitrogen™
MA556369
200 μL
£734.00
200µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

BTLA Monoclonal antibody specifically detects BTLA in Human samples. It is validated for Immunohistochemistry (Paraffin), Western Blot
Specifications

Specifications

BTLA
1 mg/mL
Monoclonal
Liquid
RUO
PBS with 50% glycerol, 0.5% BSA and 0.09% sodium azide
A630002H24; B and T lymphocyte associated; B and T lymphocyte associated protein; B and T lymphocyte attenuator; B- and T-lymphocyte attenuator; B- and T-lymphocyte-associated protein; BTLA; BTLA_HUMAN; BTLA1; CD272; CD272 antigen; FLJ16065; MGC129743
Recombinant human BTLA (proprietary range information).
Primary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
BTLA
K06312_3D7
Immunohistochemistry (Paraffin), Western Blot
Unconjugated
Mouse
Human
Q7Z6A9
151888
IgG1
Protein A
Antibody
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.