missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTLA/CD272 Mouse anti-Human, Clone: BTLA/1528, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-07803-100ug
This item is not returnable.
View return policy
Description
BTLA/CD272 Monoclonal specifically detects BTLA/CD272 in Human samples. It is validated for ELISA.
Specifications
| BTLA/CD272 | |
| Monoclonal | |
| 0.2 mg/mL | |
| ELISA | |
| B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743 | |
| Recombinant full-length human BTLA/CD272 protein | |
| 100 μg | |
| Cell Cycle and Replication, Phospho Specific | |
| 151888 | |
| Store at 4°C. | |
| IgG1 κ |
| Flow Cytometry, ELISA | |
| BTLA/1528 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction