missing translation for 'onlineSavingsMsg'
Learn More

BTLA/CD272 Antibody (MIH26), Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18390093 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18390093 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18390093 Supplier Novus Biologicals Supplier No. NBP314617AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BTLA/CD272 Monoclonal antibody specifically detects BTLA/CD272 in Human, Non-human Primate samples. It is validated for Flow Cytometry, Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTLA/CD272
Applications Flow Cytometry, Immunofluorescence
Classification Monoclonal
Clone MIH26
Conjugate Alexa Fluor 700
Dilution Flow Cytometry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743
Host Species Mouse
Immunogen Cells transfected with human BTLA/CD272
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 151888
Target Species Human, Non-human Primate
Content And Storage Store at 4°C in the dark.
Product Type Antibody
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.