missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTLA/CD272 Antibody (HMBT-6B2), PE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-81118PE
This item is not returnable.
View return policy
Description
BTLA/CD272 Monoclonal specifically detects BTLA/CD272 in Mouse samples. It is validated for Flow Cytometry.
Specifications
| BTLA/CD272 | |
| Monoclonal | |
| PE | |
| PBS | |
| BTLA | |
| This antibody was raised by immunising Armenian hamsters with mouse BTLA/CD272:Fc fusion protein. | |
| 0.1 mL | |
| Cell Cycle and Replication, Phospho Specific | |
| 151888 | |
| Mouse | |
| Purified |
| Flow Cytometry | |
| HMBT-6B2 | |
| Flow Cytometry | |
| B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| This antibody is specific for B and T lymphocyte attenuator (BTLA), which functions as a negative regulator of T cell activation and proliferation, and attenuates B cell proliferation upon associating with its known ligand, herpes virus entry mediator (HVEM). | |
| Store at 4°C in the dark. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction