missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
BTLA/CD272 Monoclonal specifically detects BTLA/CD272 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunoblotting.
Specifications
Specifications
| Antigen | BTLA/CD272 |
| Applications | Western Blot, Flow Cytometry, ELISA, Immunoblot |
| Classification | Monoclonal |
| Clone | HMBT-6B2 |
| Conjugate | Alexa Fluor 532 |
| Dilution | Western Blot, Flow Cytometry, ELISA, Immunoblotting |
| Formulation | 50mM Sodium Borate |
| Gene Alias | B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743 |
| Gene Symbols | BTLA |
| Host Species | Rabbit |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?