missing translation for 'onlineSavingsMsg'
Learn More

BTLA/CD272 Antibody (HMBT-6B2), Alexa Fluor™ 350, Novus Biologicals™

Product Code. 18395010 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18395010 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18395010 Supplier Novus Biologicals Supplier No. NBP281118AF350

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

BTLA/CD272 Monoclonal specifically detects BTLA/CD272 in Mouse samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunoblotting.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTLA/CD272
Applications Western Blot, Flow Cytometry, ELISA, Immunoblot
Classification Monoclonal
Clone HMBT-6B2
Conjugate Alexa Fluor 350
Dilution Western Blot, Flow Cytometry, ELISA, Immunoblotting
Formulation 50mM Sodium Borate
Gene Alias B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743
Gene Symbols BTLA
Host Species Rabbit
Immunogen This antibody was raised by immunising Armenian hamsters with mouse BTLA/CD272:Fc fusion protein.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication, Phospho Specific
Primary or Secondary Primary
Gene ID (Entrez) 151888
Test Specificity This antibody is specific for B and T lymphocyte attenuator (BTLA), which functions as a negative regulator of T cell activation and proliferation, and attenuates B cell proliferation upon associating with its known ligand, herpes virus entry mediator (HVEM).
Target Species Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.