missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTLA/CD272 Antibody (2161A), mFluor Violet 450 SE, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals FAB33542MFV450
372.30 GBP valid until 2025-12-16
BEST PRICE on Promo! Use promo code "24090" to get your promotional price.
This item is not returnable.
View return policy
Description
BTLA/CD272 Monoclonal antibody specifically detects BTLA/CD272 in Human samples. It is validated for Flow Cytometry, CyTOF
Specifications
| BTLA/CD272 | |
| Monoclonal | |
| mFluor Violet 450 SE | |
| 50mM Sodium Borate | |
| Rabbit | |
| 0.1 mL | |
| Cell Cycle and Replication, Phospho Specific | |
| 151888 | |
| Store at 4C in the dark. |
| Flow Cytometry, CyTOF | |
| 2161A | |
| Flow Cytometry, CyTOF-ready | |
| B and T lymphocyte associated, B and T lymphocyte attenuator, B- and T-lymphocyte attenuator, B- and T-lymphocyte-associated protein, BTLA1, CD272, CD272 antigen, FLJ16065, MGC129743 | |
| Mouse myeloma cell line, NS0-derived human BTLA, Ile25-Ser150, Accession # AAP44003 | |
| RUO | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction