missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human BTK, His Tag Recombinant Protein
BTK Recombinant Human Protein
£440.00 - £6900.00
Specifications
Accession Number | NP_000052 |
---|---|
Gene Alias | BTK, ATK, XLA |
Species | Human |
Protein Tag | His Tag |
Expression System | Baculovirus |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
12624770
|
Invitrogen™
PR5442A |
100 μg |
£1636.00
100µg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10358152
|
Invitrogen™
PV3587 |
1mg |
£6900.00
1mg |
Estimated Shipment: 16-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
11503447
|
Invitrogen™
PV3363 |
10μg |
£440.00
10µg |
Estimated Shipment: 14-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Bruton tyrosine kinase (BTK) is a cytoplasmic tyrosine kinase belonging to the SRC-related TEC subfamily of tyrosine kinases. Mutations in the BTK gene have been linked to severe developmental blocks in human B-cell ontogeny and immunodeficiency disorders.Specifications
NP_000052 | |
Human | |
Baculovirus |
BTK, ATK, XLA | |
His Tag |
For Research Use Only. Not for use in diagnostic procedures.