missing translation for 'onlineSavingsMsg'
Learn More

BTK Rabbit anti-Human, Mouse, Clone: 2J9O8, Novus Biologicals™

Product Code. 18360596 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
Unit Size:
100µL
20µL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18360596 100 μg 100µL
18328806 20 μg 20µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18360596 Supplier Novus Biologicals Supplier No. NBP315612100UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Monoclonal Antibody

BTK Monoclonal antibody specifically detects BTK in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTK
Applications Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone 2J9O8
Conjugate Unconjugated
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence 1:50 - 1:200, Immunohistochemistry-Paraffin
Formulation PBS, 0.05% BSA, 50% glycerol, pH7.3
Gene Alias Agammaglobulinaemia tyrosine kinase, AGMX1MGC126262, AT, ATKIMD1, B-cell progenitor kinase, BPK, Bruton agammaglobulinemia tyrosine kinase, Bruton tyrosine kinase, dominant-negative kinase-deficient Brutons tyrosine kinase, EC 2.7.10, EC 2.7.10.2, PSCTK1, tyrosine-protein kinase BTK, XLAMGC126261
Host Species Rabbit
Immunogen A synthetic peptide corresponding to a sequence within amino acids 560-659 of human BTK (Q06187).
Purification Method Affinity purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Apoptosis, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 695
Target Species Human, Mouse
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.