missing translation for 'onlineSavingsMsg'
Learn More

BTK IN-1, MedChemExpress

Product Code. 16463648
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16463648 10 mg 10mg
16426387 5 mg 5mg
16424736 100 mg 100mg
16400047 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16463648 Supplier Medchem Express Supplier No. HY101941

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BTK IN-1 (SNS062 analog) is a potent BTK inhibitor, with an IC50 of <100 nM.

BTK IN-1 (SNS062 analog) is a potent BTK inhibitor, with an IC50 of <100 nM.

Specifications

Chemical Name or Material BTK IN-1
CAS 1270014-40-8
Color Off-White
Purity Grade Notes Research
Molecular Formula C19H21ClN6O
Quantity 10 mg
Synonym SNS062 analog
Solubility Information DMSO : ≥ 100 mg/mL (259.83 mM)
SMILES O=C(NC1CN(C2=C3C(NC=C3)=NC=N2)CCC1)CNC4=CC=CC(Cl)=C4
Molecular Weight (g/mol) 384.86
Formula Weight 384.86
Percent Purity 98.91%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.