missing translation for 'onlineSavingsMsg'
Learn More

BTK Antibody, DyLight 488, Novus Biologicals™

Product Code. 18401216 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18401216 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18401216 Supplier Novus Biologicals Supplier No. NBP176596G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BTK Polyclonal antibody specifically detects BTK in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTK
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate DyLight 488
Formulation 50mM Sodium Borate
Gene Alias Agammaglobulinaemia tyrosine kinase, AGMX1MGC126262, AT, ATKIMD1, B-cell progenitor kinase, BPK, Bruton agammaglobulinemia tyrosine kinase, Bruton tyrosine kinase, dominant-negative kinase-deficient Brutons tyrosine kinase, EC 2.7.10, EC 2.7.10.2, PSCTK1, tyrosine-protein kinase BTK, XLAMGC126261
Host Species Rabbit
Immunogen Antibody was raised against a synthetic peptide corresponding to 16 amino acids near the amino-terminus of human BTK. The immunogen is located within amino acids 20 - 70 of BTK. Amino Acid Squence: SYYEYDFERGRRGSKK
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 695
Target Species Human, Mouse
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.