missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTK Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76596
This item is not returnable.
View return policy
Description
BTK Polyclonal specifically detects BTK in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| BTK | |
| Polyclonal | |
| Unconjugated | |
| PBS with 0.02% Sodium Azide | |
| Agammaglobulinaemia tyrosine kinase, AGMX1MGC126262, AT, ATKIMD1, B-cell progenitor kinase, BPK, Bruton agammaglobulinemia tyrosine kinase, Bruton tyrosine kinase, dominant-negative kinase-deficient Brutons tyrosine kinase, EC 2.7.10, EC 2.7.10.2, PSCTK1, tyrosine-protein kinase BTK, XLAMGC126261 | |
| Rabbit | |
| 79 kDa | |
| 0.1 mg | |
| Apoptosis, Immunology, Innate Immunity | |
| 695 | |
| Human, Mouse | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| Western Blot 0.5-2 ug/ml, ELISA 1:100-1:2000, Immunohistochemistry 5 ug/ml, Immunocytochemistry/Immunofluorescence 10 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
| Q06187 | |
| BTK | |
| Antibody was raised against a synthetic peptide corresponding to 16 amino acids near the amino-terminus of human BTK. The immunogen is located within amino acids 20 - 70 of BTK. Amino Acid Squence: SYYEYDFERGRRGSKK | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Multiple isoforms of BTK are known to exist. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction