missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTK Antibody (7F12H4), DyLight 350, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NB110-87047UV
This item is not returnable.
View return policy
Description
BTK Monoclonal specifically detects BTK in Human, Primate samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| BTK | |
| Monoclonal | |
| DyLight 350 | |
| 50mM Sodium Borate with 0.05% Sodium Azide | |
| BTK | |
| Purified recombinant C terminal fragment of BTK expressed in E. coli. [UniProt# Q06187] | |
| 0.1 mL | |
| Apoptosis, Immunology, Innate Immunity, Phospho Specific | |
| 695 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| 7F12H4 | |
| Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
| Agammaglobulinaemia tyrosine kinase, AGMX1MGC126262, AT, ATKIMD1, B-cell progenitor kinase, BPK, Bruton agammaglobulinemia tyrosine kinase, Bruton tyrosine kinase, dominant-negative kinase-deficient Brutons tyrosine kinase, EC 2.7.10, EC 2.7.10.2, PSCTK1, tyrosine-protein kinase BTK, XLAMGC126261 | |
| Mouse | |
| Ammonium sulfate precipitation | |
| RUO | |
| Primary | |
| Human, Primate | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction