missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
BTK Monoclonal antibody specifically detects BTK in Human samples. It is validated for Western Blot
Specifications
Specifications
| Antigen | BTK |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 3005A |
| Conjugate | mFluor Violet 610 SE |
| Formulation | 50mM Sodium Borate |
| Gene Alias | Agammaglobulinaemia tyrosine kinase, AGMX1MGC126262, AT, ATKIMD1, B-cell progenitor kinase, BPK, Bruton agammaglobulinemia tyrosine kinase, Bruton tyrosine kinase, dominant-negative kinase-deficient Brutons tyrosine kinase, EC 2.7.10, EC 2.7.10.2, PSCTK1, tyrosine-protein kinase BTK, XLAMGC126261 |
| Host Species | Rabbit |
| Immunogen | E. coli -derived recombinant human BTK, Glu280-Val377, Accession # Q06187 |
| Purification Method | Protein A or G purified from hybridoma culture supernatant |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?