missing translation for 'onlineSavingsMsg'
Learn More

BTG4 Antibody (1A6), Novus Biologicals™

Product Code. 18383069 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mg
Unit Size:
0.01mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18383069 0.1 mg 0.01mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18383069 Supplier Novus Biologicals Supplier No. H00054766M07

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BTG4 Monoclonal antibody specifically detects BTG4 in Human samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTG4
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 1A6
Conjugate Unconjugated
Dilution Western Blot 1:500, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation In 1x PBS, pH 7.4
Gene Accession No. AAH31045
Gene Alias B-cell translocation gene 4, BTG family member 4, MGC33003, PC3BProtein PC3b, protein BTG4
Host Species Mouse
Immunogen BTG4 (AAH31045, 1 a.a. ~ 206 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MRDEIATTVFFVTRLVKKHDKLSKQQIEDFAEKLMTILFETYRSHWHSDCPSKGQAFRCIRINNNQNKDPILERACVESNVDFSHLGLPKEMTIWVDPFEVCCRYGEKNHPFTVASFKGRWEEWELYQQISYAVSRASSDVSSGTSCDEESCSKEPRVIPKVSNPKSIYQVKSVPVLFYTFFLSNSKKNALIMKTKKQKNMERTKL
Purification Method IgG purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 54766
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.