missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTG2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA550204
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total BTG2 protein.
The protein encoded by this gene is a member of the BTG/Tob family. This family has structurally related proteins that appear to have antiproliferative properties. This encoded protein is involved in the regulation of the G1/S transition of the cell cycle.
Specifications
| BTG2 | |
| Polyclonal | |
| Unconjugated | |
| BTG2 | |
| AA959598; Agl; An; an-1; APRO1; B cell translocation gene 2, anti-proliferative; B-cell translocation gene 2; B-cell translocation gene 2, anti-proliferative; BTG anti-proliferation factor 2; BTG family member 2; BTG family, member 2; BTG2; cell surface alloantigen; Early induced gene B-cell translocation gene 2; nerve growth factor-inducible anti-proliferative; NGF-inducible anti-proliferative protein PC3; NGF-inducible anti-proliferative putative secreted protein; NGF-inducible protein TIS21; Pc3; pheochromacytoma cell-3; protein BTG2; Tis21 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12227, 29619, 7832 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1.0 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P27049, P78543, Q04211 | |
| BTG2 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-158 of human BTG family, member 2. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction