missing translation for 'onlineSavingsMsg'
Learn More

BTF3L4 Antibody (2G10), Novus Biologicals™

Product Code. 18413409 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18413409 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18413409 Supplier Novus Biologicals Supplier No. H00091408M05100ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

BTF3L4 Monoclonal antibody specifically detects BTF3L4 in Human samples. It is validated for Western Blot, ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTF3L4
Applications Western Blot, ELISA
Classification Monoclonal
Clone 2G10
Conjugate Unconjugated
Formulation In 1x PBS, pH 7.4
Gene Accession No. NP_689478.1
Gene Alias basic transcription factor 3-like 4MGC23908, MGC88389, RP4-800M22.5, transcription factor BTF3 homolog 4
Host Species Mouse
Immunogen BTF3L4 (NP_689478.1, 1 a.a. ∽ 158 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. MNQEKLAKLQAQVRIGGKGTARRKKKVVHRTATADDKKLQSSLKKLAVNNIAGIEEVNMIKDDGTVIHFNNPKVQASLSANTFAITGHAEAKPITEMLPGILSQLGADSLTSLRKLAEQFPRQVLDSKAPKPEDIDEEDDDVPDLVENFDEASKNEAN
Purification Method Protein A or G purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline DNA replication Transcription Translation and Splicing
Primary or Secondary Primary
Gene ID (Entrez) 91408
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.