missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BTF3 Recombinant Rabbit Monoclonal Antibody (K01_1N69)
GREENER_CHOICE

Product Code. 30826957
Change view
Click to view available options
Quantity:
50 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30826957 100 μL
30828155 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30826957 Supplier Invitrogen™ Supplier No. MA563324

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Recombinant Monoclonal Antibody

Immunogen sequence: RRTGAPAQAD SRGRGRARGG CPGGEATLSQ PPPRGGTRGQ EPQMKETIMN QEKLAKLQAQ VRIGGKGTAR RKKKVVHRTA TADDKKLQFS LKKLGVNNIS GIEE Highest antigen sequence identity to the following orthologs: Mouse - 89%, Pig - 97%, Cynomolgus monkey - 99%.

This gene encodes the basic transcription factor 3. This protein forms a stable complex with RNA polymerase IIB and is required for transcriptional initiation. Alternative splicing results in multiple transcript variants encoding different isoforms. This gene has multiple pseudogenes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTF3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Recombinant Monoclonal
Clone K01_1N69
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 50mM tris glycine with 40% glycerol, 0.05% BSA, 150mM NaCl and 0.01% sodium azide; pH 7.4
Gene BTF3
Gene Accession No. P20290, Q64152
Gene Alias 1700054E11Rik; basic transcription factor 3; BETA-NAC; BTF3; BTF3a; BTF3b; NACB; NAC-beta; Nascent polypeptide-associated complex subunit beta; nascent-polypeptide-associated complex beta polypeptide; OK/SW-cl.8; RNA polymerase B transcription factor 3; transcription factor BTF3
Gene Symbols BTF3
Host Species Rabbit
Immunogen Recombinant protein of human BTF3 (aa 2-105).
Purification Method Protein A/G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 218490, 294680, 689
Target Species Human, Mouse, Rat
Content And Storage Maintain refrigerated at 2-8°C for up to 1 month. For long term storage store at -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.