missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTF Antibody (M33-P5B11) - BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
£302.00
Specifications
| Antigen | BTF |
|---|---|
| Clone | M33-P5B11 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Applications | Western Blot, ELISA, Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18650517
|
Novus Biologicals
NBP2-50118-0.025ml |
0.025 mL |
N/A
|
N/A | |||||
|
18658997
|
Novus Biologicals
NBP2-50118 |
0.1 mL |
£302.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BTF Monoclonal antibody specifically detects BTF in Human samples. It is validated for Western Blot, ELISA, ImmunohistochemistrySpecifications
| BTF | |
| 1 mg/mL | |
| Western Blot, ELISA, Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Apoptosis, DNA Repair | |
| PBS | |
| 9774 | |
| IgG1 κ | |
| Protein A purified |
| M33-P5B11 | |
| Western Blot 1:100 - 1:2000, ELISA 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| bcl-2-associated transcription factor 1, BCL2-associated transcription factor 1, Btf, BTFKIAA0164bK211L9.1 | |
| Peptide Sequence - KYQGDGIVED (amino acids 900 - 909) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title