missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTD Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16330-1-AP-150UL
This item is not returnable.
View return policy
Description
Biotinidase functions to recycle biotin in the body by cleaving biocytin (biotin-epsilon-lysine), a normal product of carboxylase degradation, resulting in regeneration of free biotin. Biotinidase has also been shown to have biotinyl-transferase activity. Defects in the biotinidase gene cause multiple carboxylase deficiency.Specifications
| BTD | |
| Polyclonal | |
| Unconjugated | |
| BTD | |
| Biotinase, biotinidase, BTD | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 26363, 306262, 686 | |
| -20°C | |
| Liquid |
| Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
| 0.13 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P43251, Q5FVF9, Q8CIF4 | |
| BTD | |
| BTD Fusion Protein Ag9449 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction