missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BTD
GREENER_CHOICE

Product Code. 18705956
Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
This item is not returnable. View return policy

Product Code. 18705956

Brand: Tocris Bioscience™ 6940/10

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Selective TRPC5 activator

BTD is a selective TRPC5 activator (EC50 = 1.4 μM in a calcium influx assay, 1.3 μM in whole cell patch clamp assay). Displays >14.5-fold selectivity for TRPC5 over other TRP channels (EC50 = 20.6 μM for TRPM8). Also activates TRPC1:5 and TRPC4:5 heteromers.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 6-Methyl-N-[3-(tricyclo[3.3.1.13,7]dec-1-yloxy)propyl]-4H-1,2,4-benzothiadiazine-3-propanamide-1,1-dioxide
CAS 896684-04-1
Quantity 10 mg
Molecular Formula C24H33N3O4S
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.