missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BTBD6 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA599596
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total BTBD6.
Adapter protein for the CUL3 E3 ubiquitin-protein ligase complex. Involved in late neuronal development and muscle formation.
Specifications
| BTBD6 | |
| Polyclonal | |
| Unconjugated | |
| BTBD6 | |
| BDPL; BTB (POZ) domain containing 6; BTB domain containing 6; BTB domain protein BDPL; BTB/POZ domain-containing protein 6; Btbd6; glucocorticoid receptor AF-1 coactivator-1; lens BTB domain protein; LOW QUALITY PROTEIN: BTB/POZ domain-containing protein 6 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 690367, 90135 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q96KE9 | |
| BTBD6 | |
| A synthesized peptide derived from human BTBD6(Accession Q96KE9), corresponding to amino acid residues G296-Y346. | |
| 100 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction