missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BTBD3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568651
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%; Zebrafish: 86%.
BTBD3 is a protein containing BTB/POZ domain. Its function has not been determined yet.
Specifications
| BTBD3 | |
| Polyclonal | |
| Unconjugated | |
| BTBD3 | |
| BTB (POZ) domain containing 3; BTB domain containing 3; BTB/POZ domain containing protein 3; BTB/POZ domain-containing protein 3; BTBD3; dJ742J24.1; KIAA0952; mKIAA0952 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 228662 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| P58545 | |
| BTBD3 | |
| synthetic peptide directed towards the C-terminal region of mouse BTBD3. | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction