missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTBD3 Antibody (OTI1A10), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71699F
This item is not returnable.
View return policy
Description
BTBD3 Monoclonal antibody specifically detects BTBD3 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| BTBD3 | |
| Monoclonal | |
| FITC | |
| PBS | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| 22903 | |
| Store at 4C in the dark. | |
| IgG1 |
| Western Blot | |
| OTI1A10 | |
| Western Blot | |
| BTB (POZ) domain containing 3, BTB/POZ domain-containing protein 3, KIAA0952dJ742J24.1, MGC130038, MGC130039 | |
| Full length human recombinant protein of human BTBD3 (NP_852108) produced in E.coli. | |
| 0.1 mL | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction