missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTBD15 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78185
This item is not returnable.
View return policy
Description
BTBD15 Polyclonal specifically detects BTBD15 in Human samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| BTBD15 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| BTB (POZ) domain containing 15, BTB/POZ domain-containing protein 15, BTBD15, HSPC063, MGC57431, MGC60348, MGC88058, zinc finger and BTB domain containing 44, zinc finger and BTB domain-containing protein 44, Zinc finger protein 851, ZNF851MGC26123 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 29068 | |
| Store at 4C. Do not freeze. |
| Western Blot, Immunoprecipitation | |
| 1.0 mg/mL | |
| Western Blot 1:100-1:2000, Immunoprecipitation 2-10 mcg/mg | |
| BAI46608.1 | |
| ZBTB44 | |
| The immunogen for this product maps to a region between residue 350 and 400 of human Zinc Finger and BTB Domain Containing 44 using the numbering given in entry BAI46608.1 (GeneID 29068). | |
| 100 μL | |
| Primary | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction