missing translation for 'onlineSavingsMsg'
Learn More

BTBD15 Antibody, Alexa Fluor™ 700, Novus Biologicals™

Product Code. 18414877 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.1mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18414877 0.1 mL 0.1mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18414877 Supplier Novus Biologicals Supplier No. NBP178184AF700

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

BTBD15 Polyclonal antibody specifically detects BTBD15 in Human samples. It is validated for Western Blot, Immunohistochemistry
TRUSTED_SUSTAINABILITY

Specifications

Antigen BTBD15
Applications Western Blot, Immunohistochemistry
Classification Polyclonal
Conjugate Alexa Fluor 700
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias BTB (POZ) domain containing 15, BTB/POZ domain-containing protein 15, BTBD15, HSPC063, MGC57431, MGC60348, MGC88058, zinc finger and BTB domain containing 44, zinc finger and BTB domain-containing protein 44, Zinc finger protein 851, ZNF851MGC26123
Host Species Rabbit
Immunogen The immunogen for this product maps to a region between residue 300 and 350 of human Zinc Finger and BTB Domain Containing 44 using the numbering given in entry BAI46608.1 (GeneID 29068).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 29068
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.