missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTBD10 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-38558-25ul
This item is not returnable.
View return policy
Description
BTBD10 Polyclonal specifically detects BTBD10 in Human samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.
Specifications
| BTBD10 | |
| Polyclonal | |
| Immunohistochemistry 1:2500 - 1:5000, Immunocytochemistry/Immunofluorescence 1 - 4 μg/mL, Immunohistochemistry-Paraffin 1:2500 - 1:5000 | |
| Q9BSF8 | |
| BTBD10 | |
| This antibody was developed against a recombinant protein corresponding to amino acids: RKLHSRPRKLYKHSSTSSRIAKGGVDHTKMSLHGASGGHERSRDRRRSSDRSRDSSHERTESQLTPCIRNVTSPTRQHHVEREKDHSSSRPS | |
| 25 μL | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at-20°C long term. Avoid freeze/thaw cycles. |
| Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| BTB (broad-complex tramtrack and bric-a-brac/pox virus and zinc finger)domain-containing protein 10, BTB (POZ) domain containing 10, BTB/POZ domain-containing protein 10, GMRP1, GMRP-1, K+ channel tetramerization protein, KSARCOSIN, MGC13007 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 84280 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction