missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ BTBD1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5109456
This item is not returnable.
View return policy
Description
Immunogen sequence: CDGTANTFRV MFKEPIEILP NVCYTACATL KGPDSHYGTK GLKKVVHETP AASKTVFFFF SSPGNNNGTS IEDGQIPEII FYT.
The C-terminus of BTBD1 binds topoisomerase I. The N-terminus contains a proline-rich region and a BTB/POZ domain (broad-complex, Tramtrack and bric a brac/Pox virus and Zinc finger), both of which are typically involved in protein-protein interactions. Subcellularly, the protein localizes to cytoplasmic bodies. The C-terminus of the protein encoded by this gene binds topoisomerase I. The N-terminus contains a proline-rich region and a BTB/POZ domain (broad-complex, Tramtrack and bric a brac/Pox virus and Zinc finger), both of which are typically involved in protein-protein interactions. Subcellularly, the protein localizes to cytoplasmic bodies. Alternative splicing results in multiple transcript variants encoding different isoforms.
Specifications
| BTBD1 | |
| Polyclonal | |
| Unconjugated | |
| BTBD1 | |
| 1190005H08Rik; BTB (POZ) domain containing 1; BTB domain containing 1; BTB/POZ domain-containing protein 1; Btbd1; C15orf1; glucose signal-repressing protein; Gsrp; HCV NS5A-transactivated protein 8; hepatitis C virus NS5A-transactivated protein 8; NS5ATP8 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 83962 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 1.27 mg/mL | |
| PBS with 50% glycerol and 0.01% thimerosal; pH 7.3 | |
| P58544 | |
| BTBD1 | |
| A synthetic peptide corresponding to a sequence within amino acids 400 to the C-terminus of human BTBD1 (NP_079514.1). | |
| 100 μL | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction