missing translation for 'onlineSavingsMsg'
Learn More

BTB06584, MedChemExpress

Product Code. 16492947
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16492947 50 mg 50mg
16486888 10 mg 10mg
16469427 5 mg 5mg
16416976 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16492947 Supplier Medchem Express Supplier No. HY15877
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress BTB06584 is a selective and IF1-dependent mitochondrial F1Fo-ATPase inhibitor without compromising ATP synthesis. BTB06584 can delays ischaemic cell death.

BTB06584 is a selective and IF1-dependent mitochondrial F1Fo-ATPase inhibitor without compromising ATP synthesis. BTB06584 can delays ischaemic cell death.

Specifications

Chemical Name or Material BTB06584
CAS 219793-45-0
Color White
Purity Grade Notes Research
Molecular Formula C19H12ClNO6S
Quantity 50 mg
Solubility Information DMSO : ≥ 47 mg/mL (112.49 mM)
SMILES ClC1=CC=C(C(OC2=C([N+]([O-])=O)C=CC(S(C3=CC=CC=C3)(=O)=O)=C2)=O)C=C1
Molecular Weight (g/mol) 417.82
Formula Weight 417.82
Percent Purity 99.83%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.