missing translation for 'onlineSavingsMsg'
Learn More

BTB-1, MedChemExpress

Product Code. 16446287
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16446287 5 mg 5mg
16429976 50 mg 50mg
16414676 100 mg 100mg
16403568 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16446287 Supplier Medchem Express Supplier No. HY101770
 Restricted Item
Estimated Shipment: 10-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

MedChemExpress BTB-1 is a potent, selective and reversible mitotic motor protein Kif18A inhibitor with an IC50 of 1.69 μM.

BTB-1 is a potent, selective and reversible mitotic motor protein Kif18A inhibitor with an IC50 of 1.69 μM.

Specifications

Chemical Name or Material BTB-1
CAS 86030-08-2
Color White
Purity Grade Notes Research
Molecular Formula C12H8ClNO4S
Quantity 5 mg
Solubility Information DMSO : ≥ 100 mg/mL (335.90 mM)
SMILES O=S(C1=CC=C(Cl)C=C1[N+]([O-])=O)(C2=CC=CC=C2)=O
Molecular Weight (g/mol) 297.71
Formula Weight 297.71
Percent Purity 99.69%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.