missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BTAF1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£145.00 - £417.00
Specifications
| Antigen | BTAF1 |
|---|---|
| Dilution | Western Blot 1:500 - 1:2000, Chromatin Immunoprecipitation 5ug antibody for 10ug-15ug of Chromatin, ELISA, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells |
| Applications | ChIP Assay, ELISA, Immunoprecipitation, Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30232046
|
Novus Biologicals
NBP3-38188-20ul |
20 μL |
£145.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30226946
|
Novus Biologicals
NBP3-38188-100ul |
100 μL |
£417.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
BTAF1 Polyclonal antibody specifically detects BTAF1 in Human,Rat samples. It is validated for ChIP assay,ELISA,Immunoprecipitation,Western BlotSpecifications
| BTAF1 | |
| ChIP Assay, ELISA, Immunoprecipitation, Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Rat | |
| ATP-dependent helicase BTAF1, BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, 170kDa (Mot1homolog, S. cerevisiae), B-TFIID transcription factor-associated 170 kDa subunit, EC 3.6.1, EC 3.6.4.-, MOT1MGC138406, TAF(II)170KIAA0940, TAF-172, TAF172BTAF1 RNA polymerase II, B-TFIID transcription factor-associated, 170 kD (Mot1homolog, S. cerevisiae), TAFII170TATA-binding protein-associated factor 172, TBP-associated factor 172 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1600-1849 of human BTAF1 (NP_003963.1).,, Sequence:, LAVQNSSLHDIQHAPKLSALKQLLLDCGLGNGSTSESGTESVVAQHRILIFCQLKSMLDIVEHDLLKPHLPSVTYLRLDGSIPPGQRHSIVSRFNNDPSIDVLLLTTHVGGLGLNLTGADTVVFVEHDWNPMRDLQAMDRAHRIGQKRVVNVYRLITRGTLEEKIMGLQKFKMNIANTVISQENSSLQSMGTDQLLDLFTLDKDGKAEKADTSTSGKASMKSILENLSDLWDQEQYDSEYSLENFMHSLK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot 1:500 - 1:2000, Chromatin Immunoprecipitation 5ug antibody for 10ug-15ug of Chromatin, ELISA, Immunoprecipitation 0.5ug - 4ug antibody for 200ug - 400ug extracts of whole cells | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.3), 50% glycerol | |
| 9044 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title