missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Bsx Polyclonal Antibody

Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 93%; Pig: 100%; Rabbit: 100%; Rat: 93%.
BSX is a DNA binding protein that functions as transcriptional activator. It is essentiel for normal postnatal growth and nursing. It is also an essential factor for neuronal neuropeptide Y and agouti-related peptide function and locomotory behavior in the control of energy balance.
Specifications
Specifications
| Antigen | Bsx |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | BSX |
| Gene Accession No. | Q810B3 |
| Gene Alias | brain specific homeobox; brain-specific homeobox protein homolog; Bsx; BSX1; Bsx1a; Bsx1b; RGD1565120 |
| Gene Symbols | BSX |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?