missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent

Product Code. 11597570 Shop All Thermo Scientific Products
Change view
Click to view available options
Standard Components:
BSTFA +1% TMCS
BSTFA +10% TMCS
Quantity:
10 g
10 x 1 mL ampules
100 g
25 g
Unit Size:
10mL
Each
Pack of 10
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Standard Components Quantity Unit Size
11597570 BSTFA +1% TMCS 10 g Each
11557851 BSTFA +1% TMCS 10 x 1 mL ampules Pack of 10
11527851 BSTFA +1% TMCS 25 g Each
11517871 BSTFA +1% TMCS 100 g Each
11577841 BSTFA +10% TMCS 10 x 1 mL ampules 10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 11597570 Supplier Thermo Scientific™ Supplier No. TS38832
 Restricted Item
Estimated Shipment: 03-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Ideal for difficult-to-silylate compounds, the Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent is derivatizing compounds.

Use Thermo Scientific™ BSTFA + 1–10% TMCS Silylation Reagent for analytes that are difficult to silylate. Stronger than BSTFA alone, this catalyzed formulation is ideal for derivatizing compounds such as fatty acid amides and slightly hindered hydroxyls for GC analysis.

  • Excellent for derivatizing fatty acid amides, slightly hindered hydroxyls and other compounds
  • Catalyzed formulation is stronger than BSTFA alone
TRUSTED_SUSTAINABILITY

Specifications

For Use With GC
Standard Concentration 98 to 100, 0.1 to 1%
Storage 2°C to 8°C
Quantity 10 g
Standard Components BSTFA +1% TMCS

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.