missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ BSSP4 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA561901

Product Code. 15769701

  • £476.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: AARIPVPPAC GKPQQLNRVV GGEDSTDSEW PWIVSIQKNG THHCAGSLLT SRWV Highest antigen sequence identity to the following orthologs: Mouse - 80%, Rat - 80%.

This gene encodes a member of the trypsin family of serine proteases. The enzyme is expressed in the airways in a developmentally regulated manner. The gene is part of a cluster of serine protease genes on chromosome 16.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

BSSP4
Polyclonal
Unconjugated
PRSS22
4733401N09Rik; brain-specific serine protease 4; BSP-2; BSSP4; BSSP-4; hBSSP-4; MGC9599; prosemin; protease, serine 22; protease, serine S1 family member 22; protease, serine, 22; PRSS22; Prss26; Serine protease 22; serine protease 26; SP001LA; Tryptase epsilon; UNQ302/PRO343
Rabbit
Antigen affinity chromatography
RUO
64063
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Immunocytochemistry
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9GZN4
PRSS22
Recombinant protein corresponding to Human BSSP4. Recombinant protein control fragment (Product #RP-101091).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.