Learn More
Invitrogen™ Mouse BSSP 4 (PRSS22) ELISA Kit
Description
The Mouse BSSP 4 (PRSS22) ELISA quantitates Ms BSSP 4 in mouse serum, plasma, or cell culture medium. The assay will exclusively recognize both natural and recombinant Ms BSSP 4. Principle of the method The Mouse BSSP 4 solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Specifications
Specifications
| Accession Number | Q9ER10 |
| Assay Range | 62.5 to 4000 pg/mL |
| Assay Sensitivity | 60 pg/mL |
| Calibration Range | 60 pg/mL to 4,000 pg/mL |
| Assay | Solid-phase sandwich ELISA |
| Conjugate | HRP |
| Product Type | ELISA |
| Protein Family | Proteases, Peptidases & Inhibitors |
| Sample Type | Plasma, Serum, Supernatant |
| Specificity | This ELISA pair antibody detects mouse BSSP-4. Other species not determined. |
| Show More |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.