missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ BSJ-4-116
Description
BSJ-4-116 is a selective CDK12 PROTAC® Degrader. BSJ-4-116 selectively degrades CDK12 over a panel of other kinases and exhibits no degradation of CDK13 or cyclin K. The compound inhibits growth of T-ALL cells and sensitizes them to PARP inhibitor Olaparib (Cat. No. 7579).
PROTAC® is a registered trademark of Arvinas Operations, Inc., and is used under license.
Specifications
Specifications
| Chemical Name or Material | N-[7-[(3 R)-3-[[5-Chloro-4-[[2-[(1-methylethyl)sulfonyl]phenyl]amino]-2-pyrimidinyl]amino]-1-piperidinyl]heptyl]-2-[[2-(2,6-dioxo-3-piperidinyl)-2,3-dihydro-1,3-dioxo-1 H-isoindol-4-yl]oxy]acetamide |
| CAS | 2519823-34-6 |
| Quantity | 5 mg |
| Product Line | Tocris |
| Target | Active Degraders |
| Molecular Formula | C40H49ClN8O8S |
| Purity | 0.98 |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?