missing translation for 'onlineSavingsMsg'
Learn More

BSJ-03-123, MedChemExpress

Product Code. 16437217
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16437217

Brand: Medchem Express HY111556

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress BSJ-03-123 is a PROTAC connected by ligands for Cereblon and CDK as a potent and novel CDK6-selective small-molecule degrader.

BSJ-03-123 is a PROTAC connected by ligands for Cereblon and CDK as a potent and novel CDK6-selective small-molecule degrader.

Specifications

Chemical Name or Material BSJ-03-123
CAS 2361493-16-3
Color Yellow
Purity Grade Notes Research
Molecular Formula C47H56N10O11
Quantity 5 mg
Solubility Information DMSO : 100 mg/mL (106.72 mM; ultrasonic and adjust pH to 3 with HCl)
SMILES O=C(C(C(C)=O)=C1C)N(C2CCCC2)C3=C1C=NC(NC4=CC=C(N5CCN(CCOCCOCCOCCNC(COC6=CC=CC7=C6C(N(C8C(NC(CC8)=O)=O)C7=O)=O)=O)CC5)C=N4)=N3
Molecular Weight (g/mol) 937.01
Formula Weight 937.01
Percent Purity 98.12%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-Kinase/protease
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.