missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ BSI 201

PARP-1 inhibitor

Specifications

Chemical Name or Material 4-Iodo-3-nitrobenzamide
CAS 160003-66-7
View More Specs

Description

Description

PARP1 inhibitor. Inhibits growth of certain breast cancer cell lines in vitro. Non-selectively modifies cysteine-containing proteins in tumor cells. Inhibits ionizing radiation-induced DNA single-stranded breaks in lymphoid cell lines in vivo.
Specifications

Specifications

4-Iodo-3-nitrobenzamide
160003-66-7
Videos
Safety and Handling

Safety and Handling

Recommended Storage : Store at Room Temperature

SDS
Documents

Documents

Product Certifications
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.