Learn More
BSEP Mouse anti-Human, CoraLite 488, Clone: 3C11D5, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67512-100UL
Description
The BSEP antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human BSEP. This antibody recognizes human antigen. The BSEP antibody has been validated for the following applications: IF analysis. This antibody is coralite™488.
Specifications
| BSEP | |
| Monoclonal | |
| CoraLite 488 | |
| 100 μL | |
| 8647 | |
| IgG1 |
| Immunofluorescence | |
| 3C11D5 | |
| Mouse | |
| Primary | |
| Human |