missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BSAP/PAX5 Mouse anti-Human, Clone: 2H2D6, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 60349-1-IG-150UL
This item is not returnable.
View return policy
Description
This antibody is specific to PAX5. It will not cross react with other PAX members.
PAX5 is a member of the paired box (PAX) family of transcription factors. The PAX proteins are important regulators in early development and alterations in the expression of their genes are thought to contribute to neoplastic transformation. PAX5 is the B-cell lineage specific activator protein (BSAP) that is expressed at early, but not late stages, of B-cell differentiation. Its expression has also been detected in developing CNS and testis, therefore, PAX5 may not only play an important role in B-cell differentiation, but also in neural development and spermatogenesis. Mutations in the gene can result in leukemia, acute lymphoblastic 3.Specifications
| BSAP/PAX5 | |
| Monoclonal | |
| 1.5 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q02548 | |
| PAX5 | |
| BSAP,PAX5 Fusion Protein Ag16023 | |
| 150 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| 2H2D6 | |
| Unconjugated | |
| PAX5 | |
| BSAP, BSAP,PAX5, paired box 5, Paired box protein Pax 5, PAX5 | |
| Mouse | |
| Protein G | |
| RUO | |
| 5079 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction