Learn More
Paragon Scientific™ BS/U/M Miniature Viscometer for Transparent Liquids
Click to view available options
Viscosity Range:
0.2 to 1 cSt
1 to 5 cSt
20 to 100 cSt
3 to 15 cSt
8 to 40 cSt
Unit Size:
Each
Description
- High Quality glass capillary viscometer
- Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
Specifications
Specifications
| Viscosity Range | 0.2 to 1 cSt |
| Calibrated | No |
| Approximate Constant | 0.001 |
| ASTM Numbers | D 445, D 446 |
| Certifications/Compliance | ASTM D 445, D 446 and ISO 3104, 3105 |
| Size | M1 |
4 week lead-time