Learn More
Paragon Scientific™ BS/IP/RF U-Tube Reverse Flow for Transparent and Opaque Liquids
Product Code. 16080330
Click to view available options
Viscosity Range:
0.6 to 3 cSt
2 to 10 cSt
20 to 100 cSt
20,000 to 100,000 cSt
200 to 1000 cSt
2000 to 10,000 cSt
6 to 30 cSt
60 to 300 cSt
60,000 to 300,000 cSt
600 to 3000 cSt
6000 to 30,000 cSt
Unit Size:
Each
Product Code. 16080330
Brand: Paragon Scientific™ RF09NC
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For the determination of kinematic viscosity of transparent and opaque Newtonian liquids according to ASTM D445, ISO 3104 and IP 71 Section 1. Low viscosity aids dissipation of air bubbles which can lead to detection problems in automatic systems and optimize bath stability and uniformity.
  • High Quality glass capillary viscometer
  • Manufactured in accordance with ASTM D 445, D 446 and ISO 3104, 3105
Specifications
Viscosity Range 6000 to 30,000 cSt
Calibrated No
Approximate Constant 30
ASTM Numbers D 445, D 446
Certifications/Compliance ASTM D 445, D 446 and ISO 3104, 3105
Size 9
4 week lead-time