missing translation for 'onlineSavingsMsg'
Learn More

Brady™ BS 1710 Pipemarking App for Brady Workstation

Product Code. 30281110
Change view
Click to view available options
BS 1710 Pipemarking App
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code.
30281110 BS 1710 Pipemarking App
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30281110 Supplier Brady™ Supplier No. 149408

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Increase workplace safety and compliance with the BS 1710 Pipemarking App for Brady Workstation. Quickly and easily design British Standard-compliant pipemarkers in just a few clicks and print the self-adhesive labels with the BradyJet J5000 Colour Label Printer.

  • For Use With (Equipment): J4000, J5000

Specifications

Product Type Software
For Use With (Equipment) Brady Label Printers
Format Email with Activation Code
Software Type Sign and Label Design Software
Alternate Part Number BWS-UKPM-EM
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.