Learn More
Emma Safety Footwear Bryce XD Safety Shoes
Product Code. 15968361
Change view
Click to view available options
Size:
39
40
41
42
43
44
45
46
47
48
49
Unit Size:
1 pair
11 product options available for selection
Product selection table with 11 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size unitSize
15968361 41 1 pair
15948361 39 1 pair
15958361 40 1 pair
15978361 42 1 pair
15988361 43 1 pair
15998361 44 1 pair
15908371 45 1 pair
15918371 46 1 pair
15928371 47 1 pair
15938371 48 1 pair
15948371 49 1 pair
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 11 options available.
11 options
Product Code. 15968361 Supplier Emma Safety Footwear Supplier No. MM33186841
Only null left
Add to basket
Add to basket
The impenetrable sole protects your feet from sharp and metal objects while the steel toe cap and reinforced PU toe cover offer protection from falling objects. Microfiber construction remains flexible to stand up to cement and resists corrosive substances.
  • S3-Safety shoe with sole plate for wet work
  • GripForce™ Duo Heat Resistant Outsole
  • Microfiber construction
  • 3M™ Reflective Striping
  • Hydro-Tec™ Sanitized Silver
Specifications
Design Bryce XD
Material Microfiber
Size 41