Learn More
Emma Safety Footwear Bryce Safety Shoes
The impenetrable sole protects your feet from sharp and metal objects while the steel toe cap and reinforced PU toe cover offer protection from falling objects. Microfiber construction remains flexible to stand up to cement and resists corrosive substances.
Brand:  Emma Safety Footwear MM33084838
Additional Details : Weight : 1.52800kg
 View more versions of this product
Product Code. 15928351
  • £163.24 / 1 pair
Estimated Shipment: 27-08-2024
to see stock.
Add to basket
Description
Description
  • S3-Safety shoe with sole plate for wet work
  • GripForce™ Duo Heat Resistant Outsole
  • Microfiber construction
  • 3M™ Reflective Striping
  • Hydro-Tec™ Sanitized Silver
Specifications
Specifications
EN ISO 20345:2011
Black
Hydro-Tec™ Sanitized Silver
38
Laces
Bryce
Microfiber
Steel