Learn More
Invitrogen™ BRWD2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA588006
Description
Immunogen sequence: YFQKFQLERV NLQEVKRSTY DHTRKCTDQL LLLGQTDRAV QLLLETSADN QHYYCDSLKA CLVTTVTSSG PSQSTIKLVA TNMIANGKLA EGVQLLCLID KAADACRYLQ TYGEWNRAAW LAKVRLNPEE CADVLRRWVD HLCSPQVNQK SKALLVLLSL GCFFSVAETL HSMRYFDRAA LFVEACLKYG AFEVTEDTEK LITAIYADYA RSLKNLGFKQ GAVLFASKAG AAGKDLLNEL ESPKEEPIEE; Positive Samples: DU145, HL-60, HepG2; Cellular Location: Cytoplasm, Membrane, Nucleus, Single-pass membrane protein.
WDR11 is a member of the WD repeat protein family. WD repeats are minimally conserved regions of approximately 40 amino acids typically bracketed by gly-his and trp-asp (GH-WD), which may facilitate formation of heterotrimeric or multiprotein complexes. Members of this family are involved in a variety of cellular processes, including cell cycle progression, signal transduction, apoptosis, and gene regulation. This gene is located in the chromosome 10q25-26 region, which is frequently deleted in gliomas and tumors of other tissues, and is disrupted by the t(10;19) translocation rearrangement in glioblastoma cells. The gene location suggests that it is a candidate gene for the tumor suppressor locus.
Specifications
| BRWD2 | |
| Polyclonal | |
| Unconjugated | |
| WDR11 | |
| 2900055P10Rik; AW489876; bromodomain and WD repeat domain containing 2; bromodomain and WD repeat-containing protein 2; BRWD2; DR11; HH14; Kiaa1351; mKIAA1351; sensitization to ricin complex subunit 1; SRI1; WD repeat domain 11; WD repeat domain 15; WD repeat-containing protein 11; WD repeat-containing protein 15; WDR11; WDR15 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 55717 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 2.78 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q9BZH6 | |
| WDR11 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 975-1224 of human WDR11 (NP_0605878). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.